missing translation for 'onlineSavingsMsg'
Learn More

TTC11 Rabbit anti-Human, Mouse, Rat, Clone: 6S1G4, Novus Biologicals™

Product Code. 18375072 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375072 100 μg 100µL
18389121 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18375072 Supplier Bio-Techne Supplier No. NBP315848100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TTC11 Monoclonal antibody specifically detects TTC11 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC11
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown
Classification Monoclonal
Clone 6S1G4
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CGI-135, Fis1, FIS1 homolog, fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), fission 1 (mitochondrial outer membrane) homolog (yeast), H_NH0132A01.6, hFis1, mitochondrial fission 1 protein, tetratricopeptide repeat domain 11, Tetratricopeptide repeat protein 11, TTC11TPR repeat protein 11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-122 of human TTC11 (NP_057152.2).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Mitochondrial Fusion Proteins, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 51024
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.