missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxoid UK TTC Solution 1% 10x5mL
Description
Oxoid™ TTC Solution is used for the enumeration and identification of coliform bacteria.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Country of Origin | UK |
| Form | Liquid |
| Media Type | Supplement |
| Product Type | Solution |
| For Use With (Equipment) | Supplement for K-F Streptococcus Agar (CM0701) |
| Packaging Type | 10 x 5 mL |
| Quantity | 10 x 5mL |
| Yield | Each vial for 500mL medium |
| Species | Enterococci |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?