missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TTBK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534369
This item is not returnable.
View return policy
Description
PA5-34369 can be used with blocking peptide PEP-1412.
Tau tubulin kinase (TTBK2) is a serine/threonine kinase whose substrates are thought to include the tau and tubulin proteins. Mutations in the TTBK2 gene cause spinocerebellar ataxia type 11 (SCA11), a neurodegenerative disease characterized by progressive ataxia and atrophy of the cerebellum and brainstem. Recent studies have shown that increased levels of TTBK2 expression in kidney carcinoma and melanoma cell lines increases resistance to the chemotherapeutic agent Sunitinib, and that reduction of TTBK2 though the use of siRNA sensitized these cell lines to the effects of Sunitib, suggesting that TTBK2 can be used as a resistance marker as well a potential target to overcome resistance.
Specifications
| TTBK2 | |
| Polyclonal | |
| Unconjugated | |
| TTBK2 | |
| 2610507N02Rik; AI326283; B930008N24Rik; Bby; KIAA0847; mKIAA0847; Protein bartleby; SCA11; tau tubulin kinase 1; tau tubulin kinase 2; tau-tubulin kinase 2; Ttbk; Ttbk1; TTBK2; TTK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 146057 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q6IQ55 | |
| TTBK2 | |
| An 18 amino acid synthetic peptide near the carboxy terminus of human TTBK2. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction