missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TTBK1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12818832
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12818832 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12818832 Supplier Invitrogen™ Supplier No. PA520686

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Jurkat cell lysate. PA5-20686 can be used with blocking peptide PEP-0802.

TTBK1 is a member of the casein kinase 1 superfamily, and it functions as a neuron-specific serine/threonine and tyrosine kinase. Its primary role is to regulate the phosphorylation of tau, a protein that stabilizes microtubule assemblies. It has been found that genetic variants in this gene are linked to Alzheimer's disease. Additionally, TTBK1 is associated with several other diseases, including Childhood-Onset Schizophrenia and Spinocerebellar Ataxia 11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTBK1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TTBK1
Gene Accession No. Q5TCY1, Q6PCN3
Gene Alias AU017937; AW048023; BDTK; Brain-derived tau kinase; C330008L01Rik; KIAA1855; RP3-330M21.4; tau tubulin kinase 1; tau-tubulin kinase 1; Ttbk1
Gene Symbols TTBK1
Host Species Rabbit
Immunogen A 16 amino acid peptide near the carboxy terminus of human TTBK1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106763, 84630
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.