missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSX Universal Series General Purpose Ultra-Low Freezer

Product Code. 30694890 Shop All Thermo Scientific Products
Change view
Click to view available options
Model:
General Purpose Ultra-Low Freezer 700
Ultra-Low Freezer 400
Ultra-Low Freezer 500
Ultra-Low Freezer 600
Ultra-Low Freezer 700
Holds:
2 in. Boxes (400)
2 in. Boxes (500)
2 in. Boxes (600)
2 in. Boxes (700)
Rack Capacity:
16 Racks
20 Racks
24 Racks
28 Racks
Capacity (Metric):
549 L
682 L
816 L
949 L
950 L
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Holds Rack Capacity Capacity (Metric) Unit Size
30694890 Ultra-Low Freezer 600 2 in. Boxes (600) 24 Racks 816 L Each
18080030 General Purpose Ultra-Low Freezer 700 2 in. Boxes (700) 28 Racks 949 L Each
30694889 Ultra-Low Freezer 400 2 in. Boxes (400) 16 Racks 549 L Each
30694888 Ultra-Low Freezer 500 2 in. Boxes (500) 20 Racks 682 L Each
31935505 Ultra-Low Freezer 700 2 in. Boxes (700) 28 Racks 950 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 30694890 Supplier Thermo Scientific™ Supplier No. TSX60086FAULTS
  • ONLINE EXCLUSIVE PRICE
    £20956.51
    £12922.55 / Pack of 1
    Save £8033.96
    38% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

TSX Universal Series Ultra-Low Freezers - The next generation of our most trusted ULT platform adapts to your needs, reduces your environmental impact, and is built on our most robust design ever.

TSX Universal—Over 80 years of innovation packed into the next-gen ultra-low freezer. Recognizing the impact of your work, we prioritize sample security and reliability, and our legacy of crafting state-of-the-art lab equipment means our focus remains on user-centric design. The TSX Universal heralds a new era in performance with Universal V-Drive technology, delivering tighter control and faster recovery times. Coupled with an expanded setpoint range, universal voltage, whisper-quiet operation, simplified software updates and much more, TSX Universal seamlessly adapts to your workflow. Redefining sustainability, it is ENERGY STAR† certified with a 32% efficiency improvement, ACT† labeled, and is produced in our Zero-Waste to Landfill† certified facility. TSX Universal ULT—The pinnacle of reliability, sustainability, and innovation.

Reliability at its core:

  • Manufactured excellence: built to the highest standards at our ISO 13485:2016 facilities
  • Proven durability: 10k+ hours of intensive lab and field runtime testing to help ensure long-term reliability
  • Proven quality: each unit undergoes rigorous end-of-line testing for quality control
  • Complete transparency: shipped with factory documentation and a certification of calibration*
  • Peace of mind: backed by our 12-year warranty** for long-term confidence

Built to perform, designed to protect:

  • Enhanced performance with universal V-Drive technology: adapts to user activity to help maintain peak performance and protect what matters most—your samples.
  • Stable temperatures: less than 4°C variation helps keep samples safe anywhere in the cabinet
  • Fast recovery: industry-leading 15-minute door recovery helps minimize temperature fluctuations and downtime
  • Rapid cooling, faster setup: reaches -80°C in under 5 hours—41% faster than previous models
  • Cost effective: uses 33% less energy than previous models , helping reduce costs and environmental impact
  • Quick sample access with active pressure relief: innovative pressure relief technology enables re-entry in just 30 seconds so you can retrieve samples as frequently as needed, helping minimize disruptions
  • Maximum protection & compact footprint: advanced vacuum insulation and environmentally friendly 0 GWP, water-blown foam insulation safeguard samples while providing a compact footprint

Greener by design

  • Less hazardous: uses non-hydrofluorocarbon (HFC) refrigerants and foam insulation which has a 0 GWP and Ozone Depleting Potential (ODP) and is water-blown, helping reduce chemical emissions and outgassing
  • Less waste: contains up to 70% recycled materials, with 90% of waste diverted from landfills at our zero-waste facility***†
  • More energy efficient: ENERGY STAR® certified†, helping reduce lab energy costs and environmental footprint
  • Responsibly packaged: uses non-hydrofluorocarbon (non-HFC) refrigerants with low Global Warming Potential (GWP) and foam insulation, with 0 GWP and Ozone Depleting Potential (ODP), that is water-blown, helping reduce chemical emissions and outgassing
  • Extended life: offers a 12-year warranty, longer than any comparable product on the market

Enhanced experience, optimized for ease

  • Lab-ready convenience: universal voltage, multiple size options, and whisper-quiet operation for easy installation and compatibility
  • Reliable storage and organization: ergonomic handle helps ensure an airtight seal to prevent temperature fluctuations, while adjustable racking maximizes storage efficiency
  • Enhanced security and monitoring: 7 in. UI helps streamline operations, optional HID access enhances security, built-in power and outputs enable real-time monitoring and diagnostics, and an optional lightbar allows instant unit status visibility
  • Easy frost management: 7-point gasket design with an adjustable perimeter heater helps minimize frost build up for easier maintenance
  • Seamless integration: built-in ports for LN2, CO2, and monitoring probes helps ensure compatibility with your lab setup

*Shipped documents:

Certificate of conformance, CE declaration of conformance, certificate of calibration, QC test log sheets, and QC temperature graphs.

**Restrictions and exclusions apply. Warranty is void if the unit is not maintained as set forth in the operation.

***Zero waste defined as 90% of non-hazardous waste diverted from landfill, waste to energy and incineration.

† Information not applicable to TSX Universal ULT freezers with '-ULTS' in the part number (TSX40086FA-ULTS, TSX50086FA-ULTS, TSX60086FA-ULTS.)

*1. Data is representative of performance and not published product specifications. Results should not be referenced in developing any regulatory parameters for specific customer usage. 2. Freezer performance will vary based on customer product volume, storage format, operating conditions, test methodology and performing recommended maintenance 3. Continuous product enhancements may, without notice, result in amendments or omissions to this technical data sheet. Thermo Scientific cannot accept responsibility for damage, injury, loss or expenses resulting from misapplication of the information herein.

TRUSTED_SUSTAINABILITY

Specifications

Type Ultra-Low Freezer, Upright
Capacity (English) 28.8 cu. ft.
Capacity (Metric) 816 L
Temperature Range -40°C to -86°C
Temperature Resolution 0.1°C
Temperature (Setpoint) -80°C
Temperature Stability 0.2°C
Door Style Outer (1), Inner (2)
No. of Doors 1
Door Opening Recovery 21 min. (20°C Ambient Running at -75°C)
Cabinet Material Scratch-resistant Painted Steel
Chamber Material Cold-rolled Steel
Insulation Vacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
Interior Optional Stainless Steel
Exterior Finish Painted Cold Rolled Steel
Shelves 3 Adjustable, 1 Fixed
Capacity (English) Shelf 245 lb.
Capacity (Metric) Shelf 111 kg
Certifications/Compliance cULus, CE, UKCA, RoHS
Display Touch-Screen LCD
Casters 4 All-Direction Casters with Locks
Ports 2 Rear Access Ports
Product Line TSX Universal Series
Warranty Warranty varies by country, please contact us for details
Defrost Manual
Model Ultra-Low Freezer 600
Peak Variation at -80C 2.5°C
Water Cooled Not Available
Dimensions (DxWxH) Exterior 37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm)
Dimensions (D x W x H) Interior 28.3 x 34.4 x 51.2 in. (71.9 x 87.3 x 130.1 cm)
Depth (English) Exterior 37.6 in.
Depth (English) Interior 28.3 in.
Depth (Metric) Exterior 95.5 cm
Depth (Metric) Interior 71.9 cm
Monitoring Options Audible and Visual High and Low Temperature Alarms, Wireless
Access Security Standard Key Lock, Padlock Loop, Setpoint and Settings Security (Optional NFC Key Card Access)
On-Board Datalogging Temperature, Events, Diagnostics
Setpoint Security 2 Modes with Full Password or PIN options
Backup Systems Optional CO2 or LN2
Connectivity Thermo Scientific Instrument Connect Compatible
Smart View Compatibility Onboard accessory 5 V and POE outlets
Amperage 8.9-4.9 A
Plug Type China
Voltage 100/230 V
Electrical Requirements 100/230 V, 50/60 Hz
Frequency 50/60 Hz
Phase Single
Wattage Compressor 938 W
Energy Usage 8.32
Green Features ACT Ecolabel (Gold)
Line Voltage Indicator Yes
Voltage Compensation ±10%
Vial Capacity 60,000 x 2 mL
Compressor V-drive
Refrigerant R290 (1st stage), R170 + R290 mix (2nd stage)
Holds 2 in. Boxes (600)
Rack Capacity 24 Racks
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.