Learn More
Novus Biologicals Tsukushi/TSK Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand:  Novus Biologicals H00025987-B01P
Product Code. 18306539
  • £463.00 / 0.05mg
Description
Tsukushi/TSK Polyclonal antibody specifically detects Tsukushi/TSK in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tsukushi/TSK
Polyclonal
Western Blot 1:500
NP_056331.2
Mouse
Protein A purified
RUO
25987
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS (pH 7.4)
E2IG4E2-induced gene 4 protein, leucine rich repeat containing 54, Leucine-rich repeat-containing protein 54, LRRC54, TSKtsukushi homolog, Tsukushi, tsukushi small leucine rich proteoglycan homolog (Xenopus laevis), tsukushi, small leucine rich proteoglycan, tsukushin
LRRC54 (NP_056331.2, 1 a.a. - 353 a.a.) full-length human protein. MPWPLLLLLAVSGAQTTRPCFPGCQCEVETFGLFDSFSLTRVDCSGLGPHIMPVPIPLDTAHLDLSSNRLEMVNESVLAGPGYTTLAGLDLSHNLLTSISPTAFSRLRYLESLDLSHNGLTALPAESFTSSPLSDVNLSHNQLREVSVSAFTTHSQGRALHVDLSHNLIHRLVPHPTRAGLPAPTIQSLNLAWNRLHAVPNLRDLPLRYLSLDGNPLAVIGPGAFAGLGGLTHLSLASLQRLPELAPSGFRELPGLQVLDLSGNPKLNWAGAEVFSGLSSLQELDLSGTNLVPLPEALLLHLPALQSVSVGQDVRCRRLVREGTYPRRPGSSPKVALHCVDTRDSAARGPTIL
0.05 mg
Primary
Human
Purified