missing translation for 'onlineSavingsMsg'
Learn More

TSSK4 Antibody, R&D Systems™

Product Code. 18612198 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μL
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18612198 25 μL 25µL
18658526 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18612198 Supplier Novus Biologicals Supplier No. NBP26271325ul
  • ONLINE EXCLUSIVE PRICE
    £365.01
    £309.75 / 25µL
    Save £55.26
    15% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

TSSK4 Polyclonal specifically detects TSSK4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSSK4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias C14orf20, EC 2.7.11, EC 2.7.11.1, MGC133264, serine/threonine kinase 22E, Serine/threonine-protein kinase 22E, STK22E, Testis-specific kinase 4, testis-specific serine kinase 4, testis-specific serine/threonine-protein kinase 4, TSK-4, TSSK-4, TSSK5TSK4
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: DFGFAKMVPSNQPVGCSPSYRQVNCFSHLSQTYCGSFAYACPEILRGLPYNPFLSDTWS
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 283629
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.