missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSSK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100921
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TSSK3.
This gene encodes a kinase expressed exclusively in the testis that is thought to play a role in either germ cell differentiation or mature sperm function.
Specifications
| TSSK3 | |
| Polyclonal | |
| Unconjugated | |
| TSSK3 | |
| 1700014N07Rik; 4930594I21Rik; serine/threonine kinase 22C; serine/threonine kinase 22C (spermiogenesis associated); serine/threonine-protein kinase 22C; spermiogenesis associated; spermiogenesis associated 3; spermiogenesis associated serine/threonine kinase 22D; SPOGA3; STK22C; Stk22d; testis specific serine kinase 3; testis specific serine kinase-3; testis-specific kinase 3; testis-specific serine kinase 3; testis-specific serine/threonine kinase 22C; testis-specific serine/threonine kinase 3b; testis-specific serine/threonine-protein kinase 3; TSK3; TSK-3; Tssk3; Tssk-3; Tssk3b | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 297891, 58864, 81629 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96PN8, Q9D2E1 | |
| TSSK3 | |
| A synthesized peptide derived from human TSSK3(Accession Q96PN8), corresponding to amino acid residues M1-I51. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction