missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TSSK2 Polyclonal antibody specifically detects TSSK2 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TSSK2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | PBS (pH 7.4) |
| Gene Accession No. | NP_443732.2 |
| Gene Alias | DGSG, DiGeorge syndrome protein G, EC 2.7.11, FLJ38613, serine/threonine kinase 22B (spermiogenesis associated), Serine/threonine-protein kinase 22B, spermiogenesis associated 2, SPOGA2EC 2.7.11.1, STK22BDGS-G, testis specific serine threonine kinase 2, Testis-specific kinase 2, testis-specific serine kinase 2, testis-specific serine/threonine-protein kinase 2, TSK-2, TSSK-2 |
| Host Species | Rabbit |
| Immunogen | TSSK2 (NP_443732.2, 1 a.a. - 358 a.a.) full-length human protein. MDDATVLRKKGYIVGINLGKGSYAKVKSAYSERLKFNVAVKIIDRKKTPTDFVERFLPREMDILATVNHGSIIKTYEIFETSDGRIYIIMELGVQGDLLEFIKCQGALHEDVARKMFRQLSSAVKYCHDLDIVHRDLKCENLLLDKDFNIKLSDFGFSKRCLRDSNGRIILSKTFCGSAAYAAPEVLQSIPYQPKVYDIWSLGVILYIMVCGSMPYDDSDIRKMLRIQKEHRVDFPRSKNLTCECKDLIYRMLQPDVSQRLHIDEILSHSWLQPPKPKATSSASFKREGEGKYRAECKLDTKTDLRPDHRPDHKLGAKTQHRLLVVPENENRMEDRLAETSRAKDHHISGAEVGKAST |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?