missing translation for 'onlineSavingsMsg'
Learn More

TSSK2 Antibody (1F6), Novus Biologicals™

Product Code. 18392909 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392909 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392909 Supplier Novus Biologicals Supplier No. H00023617M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TSSK2 Monoclonal antibody specifically detects TSSK2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSSK2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1F6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH37781
Gene Alias DGSG, DiGeorge syndrome protein G, EC 2.7.11, FLJ38613, serine/threonine kinase 22B (spermiogenesis associated), Serine/threonine-protein kinase 22B, spermiogenesis associated 2, SPOGA2EC 2.7.11.1, STK22BDGS-G, testis specific serine threonine kinase 2, Testis-specific kinase 2, testis-specific serine kinase 2, testis-specific serine/threonine-protein kinase 2, TSK-2, TSSK-2
Host Species Mouse
Immunogen TSSK2 (AAH37781, 265 a.a. ~ 358 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EILSHSWLQPPKPKATSSASFKREGEGKYRAECKLDTKTDLRPDHRPDHKLGAKTQHRLLVVPENENRMEDRLAETSRAKDHHISGAEVGKAST
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 23617
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.