missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSSC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569816
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
TSSC4 is one of several tumor-suppressing subtransferable fragments located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. This gene is located among several imprinted genes; however, this gene, as well as the pan-hematopoietic expression gene (PHEMX), escapes imprinting. This gene may play a role in malignancies and disease that involve this region. Several transcript variants encoding two different isoforms have been found for this gene.
Specifications
| TSSC4 | |
| Polyclonal | |
| Unconjugated | |
| TSSC4 | |
| AA241958; ESTM671070; MNCb-3063; Protein TSSC4; Tssc4; tumor suppressing subtransferable candidate 4; tumor-suppressing STF cDNA 4 protein; tumor-suppressing subchromosomal transferable fragment 4; tumor-suppressing subchromosomal transferable fragment candidate gene 4 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10078 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y5U2 | |
| TSSC4 | |
| synthetic peptide directed towards the middle region of human TSSC4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction