missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSSC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59088
This item is not returnable.
View return policy
Description
TSSC3 Polyclonal specifically detects TSSC3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TSSC3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Beckwith-Wiedemann syndrome chromosomal region 1 candidate gene C protein, BWR1Cp17-Beckwith-Wiedemann region 1 C, HLDA2BRW1C, Imprinted in placenta and liver protein, IPLp17-BWR1C, pleckstrin homology-like domain family A member 2, pleckstrin homology-like domain, family A, member 2, TSSC3p17-Beckwith-Wiedemann region 1C, tumor suppressing subchromosomal transferable fragment cDNA 3, tumor suppressing subtransferable candidate 3, Tumor-suppressing STF cDNA 3 protein, Tumor-suppressing subchromosomal transferable fragment candidate gene 3 protein, tumor-supressing STF cDNA 3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin | |
| Q53GA4 | |
| PHLDA2 | |
| Synthetic peptides corresponding to PHLDA2(pleckstrin homology-like domain, family A, member 2) The peptide sequence was selected from the middle region of PHLDA2. Peptide sequence QNRRALQDFRSRQERTAPAAPAEDAVAAAAAAPSEPSEPSRPSPQPKPRT. | |
| 100 μL | |
| Apoptosis | |
| 7262 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction