missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSSC1 Polyclonal Antibody

TSSC1 Polyclonal Antibody
Brand: Thermo Scientific PA556685
This item is not returnable.
View return policy
Description
TSSC1 Polyclonal Antibody for Western Blot, IHC
Specifications
| TSSC1 | |
| Polyclonal | |
| Unconjugated | |
| EIPR1 | |
| EARP and GARP complex-interacting protein 1; EIPR1; Endosome-associated recycling protein-interacting protein; Golgi-associated retrograde protein-interacting protein; protein TSSC1; Tssc1; tumor suppressing subtransferable candidate 1; tumor-suppressing STF cDNA 1 protein; Tumor-suppressing subchromosomal transferable fragment candidate gene 1 protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7260 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53HC9 | |
| EIPR1 | |
| Recombinant protein corresponding to Human TSSC1. Recombinant protein control fragment (Product #RP-95958). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction