missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531806
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, H1299, HCT116, MCF-7. Predicted reactivity: Mouse (84%), Rat (85%), Rhesus Monkey (97%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be involved in 20S pre-rRNA processing.
Specifications
| TSR2 | |
| Polyclonal | |
| Unconjugated | |
| TSR2 | |
| 2310007F12Rik; DBA14; DT1P1A10; pre-rRNA-processing protein TSR2 homolog; Tsr2; TSR2 20S rRNA accumulation; TSR2, 20S rRNA accumulation, homolog; TSR2, 20S rRNA accumulation, homolog (S. cerevisiae); TSR2, ribosome maturation factor; WGG motif containing 1; WGG1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90121 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.66 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q969E8 | |
| TSR2 | |
| Recombinant protein encompassing a sequence within the center region of human TSR2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction