missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPYL6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557021
This item is not returnable.
View return policy
Description
Immunogen sequence: ATLDYALEDP HQGQRSREKS KATEVMADMF DGRLEPIVFP PPRLPEEGVA PQDPADGGHT FHILVDAGRS HGAIKAGQEV TPPP Highest antigen sequence identity to the following orthologs: Mouse - 25%, Rat - 25%.
TSPYL6 (TSPY Like 6) is a Protein Coding gene. An important paralog of this gene is TSPYL1. [GeneCards].
Specifications
| TSPYL6 | |
| Polyclonal | |
| Unconjugated | |
| TSPYL6 | |
| testicular tissue protein Li 211; testis-specific Y-encoded-like protein 6; TSPY like 6; TSPYL6; TSPY-like 6; TSPY-like protein 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 388951 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N831 | |
| TSPYL6 | |
| Recombinant protein corresponding to Human TSPYL6. Recombinant protein control fragment (Product #RP-96207). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction