missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPYL6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | TSPYL6 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
TSPYL6 Polyclonal specifically detects TSPYL6 in Human samples. It is validated for Western Blot.Specifications
| TSPYL6 | |
| Polyclonal | |
| Rabbit | |
| Q8N831 | |
| 388951 | |
| Synthetic peptides corresponding to TSPYL6(TSPY-like 6) The peptide sequence was selected from the N terminal of TSPYL6. Peptide sequence TDGSLKNGFPGEETHGLGGEKALETCGAGRSESEVIAEGKAEDVKPEECA. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| DKFZp434B125, testis-specific Y-encoded-like protein 6, TSPY-like 6, TSPY-like protein 6 | |
| TSPYL6 | |
| IgG | |
| 46 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title