missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPYL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110531
This item is not returnable.
View return policy
Description
Immunogen sequence: LKRKFIQMRR PFLERRDLII QHIPGFWVKA FLNHPRISIL INRRDEDIFR YLTNLQVQDL RHISMGYKMK LYFQTNPYFT NMVIVKEFQR NRSGRLVSHS TPIRWHRGQE PQARRHGNQD ASHSFFSWFS NHSLPEADRI AEIIKNDLWV NPLRYYLRER GSRIKRKKQE MKKRKTRGRC EVVIMEDAPD YYAVEDIFSE ISDIDETIHD IKISDFMETT D.
This gene encodes the beta-subunit of glucosidase II, an N-linked glycan-processing enzyme in the endoplasmic reticulum (ER). This protein is an acidic phospho-protein known to be a substrate for protein kinase C. Mutations in this gene have been associated with the autosomal dominant polycystic liver disease (PCLD). Alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
| TSPYL2 | |
| Polyclonal | |
| Unconjugated | |
| TSPYL2 | |
| CASK interacting nucleosome assembly protein; CASK-interacting nucleosome assembly protein; CDA1; Cell division autoantigen 1; cell division autoantigen 1 nucleolar protein; CINAP; CTCL; CTCL tumor antigen se20-4; CTCL-associated antigen se20-4; Cutaneous T-cell lymphoma-associated antigen se20-4; cutaneous T-cell lymphoma-associated tumor antigen se20-4; DENTT; differentially expressed nucleolar TGF-beta1 target; differentially-expressed nucleolar TGF-beta1 target protein; DXBwg1396e; DXHXS1008E; E130307F10Rik; HRIHFB2216; NP79; nuclear protein of 79 kDa; nucleolar TGF-beta1 target protein; RGD1563283; RP1-290F12.2; SE204; testis-specific protein Y encoded-like 2; testis-specific Y-encoded-like protein 2; TSPX; TSPY like 2; Tspyl2; TSPY-like 2; TSPY-like protein 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 302612, 52808, 64061 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.71 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q7TQI8, Q9H2G4 | |
| TSPYL2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 240-460 of human TSPYL2 (NP_071400.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction