missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPYL1 Monoclonal Antibody (4F11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA520160
This item is not returnable.
View return policy
Description
Peptide Sequence: MSGLDGVKRT TPLQTHSIII SDQVPSDQDA HQYLRLRDQS EATQVMAEPG EGGSETVALP PPPPSEEGGV PQDAAGRGGT PQIRVVGGRG HVAIKAGQEE.
The protein encoded by this gene is found in the nucleolus and is similar to that of a family of genes on the Y-chromosome. This gene is intronless. Defects in this gene are a cause of sudden infant death with dysgenesis of the testes syndrome (SIDDT).
Specifications
| TSPYL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9H0U9 | |
| TSPYL1 | |
| TSPYL1 (NP_003300.1, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 4F11 | |
| Unconjugated | |
| TSPYL1 | |
| testis-specific protein, Y-encoded-like 1; testis-specific Y-encoded-like protein 1; TSPY like 1; Tspyl; Tspyl1; TSPY-like 1; TSPY-like protein 1; Y encoded | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7259 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction