missing translation for 'onlineSavingsMsg'
Learn More

TSPO/PBR Mouse Monoclonal Antibody, Proteintech

Product Code. 30233194 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30233194 150 μL 150µL
30236079 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30233194 Supplier Proteintech Supplier No. 681371IG150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The TSPO/PBR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TSPO/PBR. This antibody recognizes human, mouse, rat antigen. The TSPO/PBR antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.

Specifications

Antigen TSPO/PBR
Applications Western Blot, Immunocytochemistry/Immunofluorescence, ELISA
Classification Monoclonal
Clone 1D1F12
Concentration 1000 μg/mL
Conjugate Unconjugated
Gene Symbols BZRP; DBI; IBP; MBR; PBR; PKBS; PTBR; mDRC; pk18
Host Species Mouse
Immunogen Recombinant Protein
Purification Method Caprylic acid/ammonium sulfate precipitation
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 706
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 18 kDa
Product Type Monoclonal Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.