missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA553110
This item is not returnable.
View return policy
Description
Immunogen sequence: LLLYHTENNV GLKNAWNIIQ AEMRCCGVTD YTDWYPVLGE NTVPDRCCME NSQGCGRNAT TPLWRTGCYE KVKMWFDD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
TSPAN9 includes the tetraspan family protein members which are characterized by four predicted transmembrane domains, and are thought to be involved in physiological processes such as tissue differentiation, immunological responses, and sperm-egg fusion. TSPAN9 has recently been identified as a platelet tetraspanin and a component of tetraspanin microdomains that include the collagen receptor GPVI (glycoprotein VI) and integrin alpha6beta1 but not the von Willebrand receptor GPIbalpha or the integrins alphaIIbbeta3 or alpha2beta1, suggesting that TSPAN9 may act to regulate platelet function in concert with other tetraspanins and their associated proteins.
Specifications
| TSPAN9 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN9 | |
| 6720474K14Rik; 9430079M16Rik; AU018597; NET5; NET-5; new EST tetraspan 5; PP1057; RGD1304740; tetraspan NET-5; tetraspanin 9; tetraspanin-9; transmembrane 4 superfamily member tetraspan NET-5; Tspan9; tspan-9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10867 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry, Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75954 | |
| TSPAN9 | |
| Recombinant protein corresponding to Human TSPAN9 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction