missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-88505
This item is not returnable.
View return policy
Description
TSPAN9 Polyclonal specifically detects TSPAN9 in Human samples. It is validated for Western Blot.
Specifications
| TSPAN9 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10867 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NET-5, NET5tetraspanin-9, new EST tetraspan 5, PP1057, Tetraspan NET-5, tetraspanin 9, transmembrane 4 superfamily member tetraspan NET-5, tspan-9 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of human TSPAN9 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction