missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TSPAN8/TM4SF3 Antibody (INSERM patent anti-CO-029) - Humanized, Novus Biologicals™

Human Monoclonal Antibody

£162.00 - £695.00

Specifications

Antigen TSPAN8/TM4SF3
Clone INSERM patent anti-CO-029
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30048191
View Documents
Novus Biologicals™
NBP3-28263-100ug
100 μg
£257.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30046405
View Documents
Novus Biologicals™
NBP3-28263-50ug
50 μg
£162.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30046126
View Documents
Novus Biologicals™
NBP3-28263-1mg
1 mg
£695.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TSPAN8/TM4SF3 Monoclonal antibody specifically detects TSPAN8/TM4SF3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

TSPAN8/TM4SF3
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Cancer
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CO-029, tetraspanin 8, TM4SF3tspan-8, Transmembrane 4 superfamily member 3tetraspanin-8, Tspan-8, Tumor-associated antigen CO-029
TSPAN8
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
INSERM patent anti-CO-029
Monoclonal
Purified
RUO
Human
P19075
7103
IgG1
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.