missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN8/TM4SF3 Antibody (INSERM patent anti-CO-029), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28263CL1
This item is not returnable.
View return policy
Description
TSPAN8/TM4SF3 Monoclonal antibody specifically detects TSPAN8/TM4SF3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| TSPAN8/TM4SF3 | |
| Monoclonal | |
| CoraFluor 1 | |
| CO-029, tetraspanin 8, TM4SF3tspan-8, Transmembrane 4 superfamily member 3tetraspanin-8, Tspan-8, Tumor-associated antigen CO-029 | |
| TSPAN8 | |
| 0.1 mL | |
| Cancer | |
| 7103 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| INSERM patent anti-CO-029 | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction