missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPAN8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599214
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TSPAN8.
TSPAN8 (tetraspanin 8), also known as TM4SF3 or CO-029, is a 237 amino acid multi-pass membrane protein that belongs to the tetraspanin family of transmembrane proteins and is thought to play a role in signal transduction events that influence cell development, activation, growth and motility. Localized to the cell membrane, TSPAN8 is expressed in colon, gastric, renal and pancreatic cancers, strongly suggesting a role in cancer formation and metastasis. The gene encoding TSPAN8 maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome.
Specifications
| TSPAN8 | |
| Polyclonal | |
| Unconjugated | |
| Tspan8 | |
| C76990; CO-029; CO-29; D6.1A; E330007O21Rik; tetraspanin 8; Tetraspanin8; tetraspanin-8; Tm4sf3; transmembrane 4 superfamily member 3; TSPAN8; Tspan-8; Tumor-associated antigen CO-029 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7103 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P19075 | |
| Tspan8 | |
| A synthesized peptide derived from human TSPAN8(Accession P19075), corresponding to amino acid residues F109-A159. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction