Learn More
Invitrogen™ TSPAN8 Biosimilar Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA559434
Descrizione
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term INSERM patent anti-CO-029.
TSPAN8 (tetraspanin 8), also known as TM4SF3 or CO-029, is a 237 amino acid multi-pass membrane protein that belongs to the tetraspanin family of transmembrane proteins and is thought to play a role in signal transduction events that influence cell development, activation, growth and motility. Localized to the cell membrane, TSPAN8 is expressed in colon, gastric, renal and pancreatic cancers, strongly suggesting a role in cancer formation and metastasis. The gene encoding TSPAN8 maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome.
Specifica
| TSPAN8 Biosimilar | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.