missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN7/TM4SF2 Antibody (B2D), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-75971H
This item is not returnable.
View return policy
Description
TSPAN7/TM4SF2 Monoclonal specifically detects TSPAN7/TM4SF2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TSPAN7/TM4SF2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TSPAN7 | |
| Human T-ALL cell line THP-6 | |
| 0.1 mL | |
| Primary | |
| The antibody QBEnd-10 reacts with Class II epitope on CD34 (Mucosialin), a 110-115 kDa monomeric transmembrane phosphoglycoprotein expressed on hematopoietic progenitors cells and on the most pluripotential stem cells; it is gradually lost on progenitor cells. This antibody has been also used as an endothelial marker. | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| B2D | |
| Western Blot, Immunohistochemistry | |
| A15CCG-B7, CD231, CD231 antigen, Cell surface glycoprotein A15, Membrane component chromosome X surface marker 1, mental retardation, X-linked 58, MRX58, MXS1T-cell acute lymphoblastic leukemia associated antigen 1, T-cell acute lymphoblastic leukemia-associated antigen 1, tetraspanin 7, tetraspanin-7, TM4SF2tetraspanin protein, transmembrane 4 superfamily 2b, Transmembrane 4 superfamily member 2X chromosome, surface marker 1, transmembrane protein A15, Tspan-7 | |
| Mouse | |
| Protein A purified | |
| metabolism | |
| 7102 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction