missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31685
This item is not returnable.
View return policy
Description
TSPAN6 Polyclonal specifically detects TSPAN6 in Human samples. It is validated for Western Blot.
Specifications
| TSPAN6 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| Putative NF-kappa-B-activating protein 321, T245, T245 protein, tetraspanin 6, Tetraspanin TM4-D, tetraspanin-6, TM4SF6tetraspan TM4SF, Transmembrane 4 superfamily member 6A15 homolog, TSPAN-6 | |
| Rabbit | |
| 28 kDa | |
| 100 μL | |
| Signal Transduction | |
| 7105 | |
| Human | |
| IgG |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| O43657 | |
| TSPAN6 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human TSPAN6. The exact sequence is proprietary. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Mouse (85%), Rat (85%), and Pig (85%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction