Learn More
TSPAN5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12122-1-AP-20UL
 View more versions of this product
Product Code. 16879643
  • £135.00 / 20µL
Description
TSPAN5 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.
Specifications
Specifications
TSPAN5
Polyclonal
Unconjugated
TSPAN5
NET 4, NET4, Tetraspan NET 4, tetraspanin 5, TM4SF9, TSPAN 5, TSPAN5
Rabbit
Antigen Affinity Chromatography
RUO
10098, 362048, 56224
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62079, P62080, Q68VK5
Tspan5
TSPAN5 Fusion Protein Ag2765
20 μL
Primary
Human, Mouse, Rat