missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN33 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14329-1-AP-150UL
This item is not returnable.
View return policy
Description
TSPAN33 plays an important role in normal erythropoiesis. It has a role in the differentiation of erythroid progenitors.Specifications
| TSPAN33 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN33 | |
| hPen, PEN, Penumbra, Proerythroblast new membrane, tetraspanin 33, Tspan 33, TSPAN33 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 232670, 340348, 500065 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86UF1, Q8R3S2 | |
| TSPAN33 | |
| TSPAN33 Fusion Protein Ag5597 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction