missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPAN33 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526404
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine and mouse based on sequence homology.
TSPAN33 plays an important role in normal erythropoiesis. It has a role in the differentiation of erythroid progenitors.
Specifications
| TSPAN33 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN33 | |
| 1300010A20Rik; hPen; hypothetical protein LOC539284; mPen; PEN; PEN.; Penumbra; proerythroblast new membrane; RGD1560915; tetraspanin 33; tetraspanin-33; Tetraspanin-33-like protein; Tspan33; tspan-33 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 340348 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q86UF1 | |
| TSPAN33 | |
| KLH conjugated synthetic peptide between 254-283 amino acids from the C-terminal region of human TSPAN33. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction