missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPAN33 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554186
This item is not returnable.
View return policy
Description
Immunogen sequence: NAIVHYRDDL DLQNLIDFGQ KKFSCCGGIS YKDWSQNMYF NCSEDNPSRE RCSVPYSCCL PTPDQAVINT MCGQGMQAFD YLEASKVIYT NGCIDKLVNW IHSNLFL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
TSPAN33 plays an important role in normal erythropoiesis. It has a role in the differentiation of erythroid progenitors.
Specifications
| TSPAN33 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN33 | |
| 1300010A20Rik; hPen; hypothetical protein LOC539284; mPen; PEN; PEN.; Penumbra; proerythroblast new membrane; RGD1560915; tetraspanin 33; tetraspanin-33; Tetraspanin-33-like protein; Tspan33; tspan-33 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 340348 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86UF1 | |
| TSPAN33 | |
| Recombinant protein corresponding to Human TSPAN33. Recombinant protein control fragment (Product #RP-91166). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction