missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN33 Antibody (545422) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8405I
This item is not returnable.
View return policy
Description
TSPAN33 Monoclonal antibody specifically detects TSPAN33 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| TSPAN33 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 340348 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 545422 | |
| Flow Cytometry, CyTOF-ready | |
| hPen, proerythroblast new membrane, tetraspanin 33, tetraspanin-33, tspan-33 | |
| NS0 mouse myeloma cell line transfected with human TSPAN33, Met1-Tyr283, Accession # NP_848657 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction