missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TSPAN32/TSSC6 Antibody (2G12), Novus Biologicals™

Product Code. 18345119 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345119 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345119 Supplier Novus Biologicals™ Supplier No. H00010077M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TSPAN32/TSSC6 Monoclonal antibody specifically detects TSPAN32/TSSC6 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN32/TSSC6
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G12
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Alias ART1, FLJ17158, FLJ97586, MGC22455, pan-hematopoietic expression, PHEMXPHMX, Protein Phemx, tetraspanin 32, tetraspanin-32, tspan-32, TSSC6pan-hematopoietic expression protein, tumor-suppressing STF cDNA 6, tumor-suppressing subchromosomal transferable fragment cDNA 6, tumor-suppressing subtransferable candidate 6
Host Species Mouse
Immunogen TSPAN32 (NP_005696, 194 a.a. ∽ 290 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RCGCSLDRKGKYTLTPRACGRQPQEPSLLRCSQGGPTHCLHSEAVAIGPRGCSGSLRWLQESDAAPLPLSCHLAAHRALQGRSRGGLSGCPERGLSD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10077
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.