Learn More
Invitrogen™ TSPAN31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139789
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DVINASWWVM SNKTRDELER SFDCCGLFNL TTLYQQDYDF CTAICKSQSP TCQMCGEKF Highest antigen sequence indentity to the following orthologs: Rat - 71%, Mouse - 75%.
TSPAN31 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is thought to be involved in growth-related cellular processes. It is associated with tumorigenesis and osteosarcoma.
Specifications
| TSPAN31 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN31 | |
| 2700085A14Rik; AW742554; sarcoma amplified sequence; sarcoma-amplified sequence; Sarcoma-amplified sequence homolog; SAS; tetraspanin 31; tetraspanin-31; transmembrane 4 protein; transmembrane protein; TSPAN31; Tspan-31; Unknown (protein for MGC:133819) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6302 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q12999 | |
| TSPAN31 | |
| Recombinant protein corresponding to Human TSPAN31. Recombinant protein control fragment (Product #RP-95638). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.