missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPAN31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569422
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 86%; Rat: 93%.
TSPAN31 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is thought to be involved in growth-related cellular processes. It is associated with tumorigenesis and osteosarcoma.
Specifications
| TSPAN31 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN31 | |
| 2700085A14Rik; AW742554; sarcoma amplified sequence; sarcoma-amplified sequence; Sarcoma-amplified sequence homolog; SAS; tetraspanin 31; tetraspanin-31; transmembrane 4 protein; transmembrane protein; TSPAN31; Tspan-31; Unknown (protein for MGC:133819) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6302 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q12999 | |
| TSPAN31 | |
| synthetic peptide directed towards the middle region of human TSPAN31. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction