missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93911-0.02ml
This item is not returnable.
View return policy
Description
TSPAN3 Polyclonal antibody specifically detects TSPAN3 in Mouse samples. It is validated for Western Blot
Specifications
| TSPAN3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| tetraspan TM4SF, tetraspanin 3, Tetraspanin TM4-A, tetraspanin-3, TM4-A, TM4SF8, Transmembrane 4 superfamily member 8tetraspan 3, TSPAN-3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 105-210 of human TSPAN3 (NP_005715.1). YVYRAKVENEVDRSIQKVYKTYNGTNPDAASRAIDYVQRQLHCCGIHNYSDWENTDWFKETKNQSVPLSCCRETASNCNGSLAHPSDLYAEGCEALVVKKLQEIMM | |
| 0.02 mL | |
| Cell Biology | |
| 10099 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction