missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSPAN17 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA565066

Product Code. 15987834

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: NDWNLNIYFNC TDLNPSRERC GVPFSCCVRD PAEDVLNTQC GYDVRLKLEL EQQGFIHTKG CVGQFEKWL Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.

Regulates ADAM10 maturation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TSPAN17
Polyclonal
Unconjugated
Tspan17
2210021G21Rik; AI047581; F-box only protein 23; F-box only protein 23, transmembrane 4 superfamily member 17; FBX23; Fbxo23; fc49c03; gamma-hydroxybutyrate receptor; GHB-R; tetraspan protein SB134; tetraspanin 17; tetraspanin-17; TM4SF17; transmembrane 4 superfamily member 17; transmembrane 4 superfamily member 17 isoform b; TSPAN17; tspan-17; wu:fc49c03; zgc:158284
Rabbit
Antigen affinity chromatography
RUO
26262
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96FV3
Tspan17
Recombinant Human TSPAN17. Recombinant protein control fragment (Product #RP-105829).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.