missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN17 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94756-0.1ml
This item is not returnable.
View return policy
Description
TSPAN17 Polyclonal antibody specifically detects TSPAN17 in Mouse samples. It is validated for Western Blot
Specifications
| TSPAN17 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| F-box only protein 23, FBXO23, MGC14859, MGC71255, Tetraspan protein SB134, tetraspanin 17, tetraspanin-17, TM4SF17, transmembrane 4 superfamily member 17, Tspan-17 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 116-234 of human TSPAN17 (NP_001006617.2). VFKDWIRDQLNLFINNNVKAYRDDIDLQNLIDFAQEYWSCCGARGPNDWNLNIYFNCTDLNPSRERCGVPFSCCVRDPAEDVLNTQCGYDVRLKLELEQQGFIHTKGCVGQFEKWLQDN | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| 26262 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction