Learn More
Invitrogen™ TSPAN16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140286
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: HTYPRSCCKS IGSVSCDGRD VSPNVIHQKG CFHKLLKITK TQS Highest antigen sequence indentity to the following orthologs: Mouse - 31%, Rat - 33%.
Specifications
| TSPAN16 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN16 | |
| tetraspanin 16; tetraspanin TM4-B; Tetraspanin-16; TM4-B; TM4SF16; TM-8; transmembrane 4 superfamily member 16; TSPAN16; Tspan-16 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26526 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UKR8 | |
| TSPAN16 | |
| Recombinant protein corresponding to Human TSPAN16. Recombinant protein control fragment (Product #RP-109567). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.