missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPAN15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542948
This item is not returnable.
View return policy
Description
Peptide sequence: LGILLPQFLG VLLTLLYITR VEDIIMEHSV TDGLLGPGAK PSVEAAGTGC Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
TSPAN15 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene. The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene.
Specifications
| TSPAN15 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN15 | |
| 1110036D12Rik; 2700063A19Rik; AW048364; NET7; NET-7; Tetraspan NET-7; tetraspanin 15; tetraspanin-15; TM4SF15; transmembrane 4 superfamily member 15; transmembrane 4 superfamily member tetraspan NET-7; Tspan15; tspan-15; UNQ677/PRO1311 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23555 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O95858 | |
| TSPAN15 | |
| Synthetic peptide directed towards the C-terminal of human TSPAN15 (aa 239-288). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction