missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPAN15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139906
This item is not returnable.
View return policy
Beskrivelse
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RVEDIIMEHS VTDGLLGPGA KPSVEAAGTG CCLCYPN Highest antigen sequence indentity to the following orthologs: Rat - 84%, Mouse - 81%.
TSPAN15 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene. The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene.
Tekniske data
| TSPAN15 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN15 | |
| 1110036D12Rik; 2700063A19Rik; AW048364; NET7; NET-7; Tetraspan NET-7; tetraspanin 15; tetraspanin-15; TM4SF15; transmembrane 4 superfamily member 15; transmembrane 4 superfamily member tetraspan NET-7; Tspan15; tspan-15; UNQ677/PRO1311 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23555 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95858 | |
| TSPAN15 | |
| Recombinant protein corresponding to Human TSPAN15. Recombinant protein control fragment (Product #RP-109252). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion