Learn More
Invitrogen™ TSPAN12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580195
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela cell, human SW620 cell, rat kidney tissue, rat brain tissue, mouse brain tissue. IHC: human lung cancer tissue, mouse lung tissue, rat lung tissue, human placenta tissue, human rectal cancer tissue.
TSPAN12 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.
Specifications
| TSPAN12 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN12 | |
| 9030619E17; AI426782; AI663988; AW111457; EVR5; MGC137611 protein; NET2; NET-2; Tetraspan NET-2; tetraspanin 12; tetraspanin-12; Tm4sf12; transmembrane 4 superfamily member 12; TSPAN12; Tspan-12; UNQ774/PRO1568; zgc:63880 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23554, 269831, 362326 | |
| -20°C | |
| Lyophilized |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| O95859, Q569A2, Q8BKT6 | |
| TSPAN12 | |
| E. coli-derived human TSPAN12 recombinant protein (Position: G111-R224). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.