missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN12 Antibody (921938) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8910L
This item is not returnable.
View return policy
Description
TSPAN12 Monoclonal antibody specifically detects TSPAN12 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| TSPAN12 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23554 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 921938 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, NET-2, ten-eleven translocation-1, tetraspanin-12 | |
| NS0 mouse myeloma cell line transfected with human TSPAN12, Met1-Leu305 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction