missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16058-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.Specifications
| TSPAN1 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN1 | |
| 9030418M05Rik, NET 1, RP11 322N21.1, Tetraspan NET 1, tetraspanin 1, Tetraspanin TM4 C, TSPAN 1, TSPAN1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10103, 298436, 66805 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60635, Q6AYR9, Q99J59 | |
| Tspan1 | |
| TSPAN1 Fusion Protein Ag8786 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction