missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN1 Rabbit anti-Human, Clone: 2022-04-12 00:00:00, Novus Biologicals™
Rabbit Monoclonal Antibody
£200.00
Specifications
| Antigen | TSPAN1 |
|---|---|
| Clone | 2022-04-12 00:00:00 |
| Concentration | 1 mg/ml |
| Dilution | ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Applications | ELISA, Immunohistochemistry, Immunofluorescence |
Description
TSPAN1 Monoclonal antibody specifically detects TSPAN1 in Human samples. It is validated for ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
| TSPAN1 | |
| 1 mg/ml | |
| ELISA, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| NET1, NET-1, tetraspan 1, Tetraspan NET-1, tetraspanin 1, Tetraspanin TM4-C, tetraspanin-1, TM4C, TM4SF, TSPAN-1 | |
| This antibody was raised by immunising BALB/c mice with the major extracellular domain of tetraspanin 1. | |
| Primary | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
| 2022-04-12 00:00:00 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 10103 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title