missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN1 Mouse anti-Human, Clone: 2022-04-12 00:00:00, Novus Biologicals™
Mouse Monoclonal Antibody
£200.00 - £463.00
Specifications
| Antigen | TSPAN1 |
|---|---|
| Clone | 2022-04-12 00:00:00 |
| Dilution | ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18328502
|
Novus Biologicals
NBP3-12060-0.025MG |
0.025 mg |
£200.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18335013
|
Novus Biologicals
NBP3-12060-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TSPAN1 Monoclonal antibody specifically detects TSPAN1 in Human samples. It is validated for ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
| TSPAN1 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS | |
| 10103 | |
| IgG1 κ | |
| Protein A purified |
| 2022-04-12 00:00:00 | |
| Monoclonal | |
| Mouse | |
| Human | |
| NET1, NET-1, tetraspan 1, Tetraspan NET-1, tetraspanin 1, Tetraspanin TM4-C, tetraspanin-1, TM4C, TM4SF, TSPAN-1 | |
| This antibody was raised by immunising BALB/c mice with the major extracellular domain of tetraspanin 1. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title