missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN1 Mouse anti-Human, Clone: 2022-04-12 00:00:00, Novus Biologicals™
Shop All Bio Techne ProductsDescription
TSPAN1 Monoclonal antibody specifically detects TSPAN1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence
Specifications
Specifications
| Antigen | TSPAN1 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | 2022-04-12 00:00:00 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | PBS |
| Gene Alias | NET1, NET-1, tetraspan 1, Tetraspan NET-1, tetraspanin 1, Tetraspanin TM4-C, tetraspanin-1, TM4C, TM4SF, TSPAN-1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?