missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN1 Antibody (010), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90167PCP
This item is not returnable.
View return policy
Description
TSPAN1 Monoclonal specifically detects TSPAN1 in Human samples. It is validated for Flow Cytometry.
Specifications
| TSPAN1 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 10103 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry | |
| 10 | |
| Flow Cytometry | |
| NET1, NET-1, tetraspan 1, Tetraspan NET-1, tetraspanin 1, Tetraspanin TM4-C, tetraspanin-1, TM4C, TM4SF, TSPAN-1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human TSPAN1 (O60635; Tyr110-Asn211). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction