missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TSPAN1 Monoclonal specifically detects TSPAN1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | TSPAN1 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 10 |
| Conjugate | DyLight 488 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | NET1, NET-1, tetraspan 1, Tetraspan NET-1, tetraspanin 1, Tetraspanin TM4-C, tetraspanin-1, TM4C, TM4SF, TSPAN-1 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human TSPAN1 (O60635; Tyr110-Asn211). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?