missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TSP50 Antibody, R&D Systems™

Rat Monoclonal Antibody
£146.00 - £458.00
Specifications
| Antigen | TSP50 |
|---|---|
| Clone | 260514 |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
18702178
![]() |
R&D Systems
MAB2455-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18121461
![]() |
R&D Systems
MAB2455 |
500 μg |
£458.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
TSP50 Monoclonal specifically detects TSP50 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Especificaciones
| TSP50 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cancer/testis antigen 20, CT20probable threonine protease PRSS50, EC 3.4.25.-, protease, serine, 50, Serine protease 50, testes-specific protease 50, Testis-specific protease-like protein 50, TSP50testes specific protease 50 | |
| Prss50 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 260514 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rat | |
| Mouse | |
| NP_666339.2 | |
| 29122 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TSP50 His58-Leu439 Accession # NP_666339.2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse TSP50 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human TSP50 is observed. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto